Merck
  • Resilient yet entirely degradable gelatin-based biogels for soft robots and electronics.
Resilient yet entirely degradable gelatin-based biogels for soft robots and electronics.
Nature materials (2020-06-17)
Melanie Baumgartner, Florian Hartmann, Michael Drack, David Preninger, Daniela Wirthl, Robert Gerstmayr, Lukas Lehner, Guoyong Mao, Roland Pruckner, Stepan Demchyshyn, Lisa Reiter, Moritz Strobel, Thomas Stockinger, David Schiller, Susanne Kimeswenger, Florian Greibich, Gerda Buchberger, Elke Bradt, Sabine Hild, Siegfried Bauer, Martin Kaltenbrunner
ABSTRACT
Biodegradable and biocompatible elastic materials for soft robotics, tissue engineering or stretchable electronics with good mechanical properties, tunability, modifiability or healing properties drive technological advance, and yet they are not durable under ambient conditions and do not combine all the attributes in a single platform. We have developed a versatile gelatin-based biogel, which is highly resilient with outstanding elastic characteristics, yet degrades fully when disposed. It self-adheres, is rapidly healable and derived entirely from natural and food-safe constituents. We merge all the favourable attributes in one material that is easy to reproduce and scalable, and has a low-cost production under ambient conditions. This biogel is a step towards durable, life-like soft robotic and electronic systems that are sustainable and closely mimic their natural antetypes.
Sigma-Aldrich
S-(−)-Eticlopride hydrochloride, ≥98% (HPLC), powder
Pricing and availability is not currently available.