Merck
  • Cryo-EM structures from sub-nl volumes using pin-printing and jet vitrification.
Cryo-EM structures from sub-nl volumes using pin-printing and jet vitrification.
Nature communications (2020-05-24)
Raimond B G Ravelli, Frank J T Nijpels, Rene J M Henderikx, Giulia Weissenberger, Sanne Thewessem, Abril Gijsbers, Bart W A M M Beulen, Carmen López-Iglesias, Peter J Peters
ABSTRACT
The increasing demand for cryo-electron microscopy (cryo-EM) reveals drawbacks in current sample preparation protocols, such as sample waste and lack of reproducibility. Here, we present several technical developments that provide efficient sample preparation for cryo-EM studies. Pin printing substantially reduces sample waste by depositing only a sub-nanoliter volume of sample on the carrier surface. Sample evaporation is mitigated by dewpoint control feedback loops. The deposited sample is vitrified by jets of cryogen followed by submersion into a cryogen bath. Because the cryogen jets cool the sample from the center, premounted autogrids can be used and loaded directly into automated cryo-EMs. We integrated these steps into a single device, named VitroJet. The device's performance was validated by resolving four standard proteins (apoferritin, GroEL, worm hemoglobin, beta-galactosidase) to ~3 Å resolution using a 200-kV electron microscope. The VitroJet offers a promising solution for improved automated sample preparation in cryo-EM studies.
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VIII, lyophilized powder, ≥500 units/mg protein