Merck
  • Immunohistochemical expression of P-selectin, SP-A, HSP70, aquaporin 5, and fibronectin in saltwater drowning and freshwater drowning.
Immunohistochemical expression of P-selectin, SP-A, HSP70, aquaporin 5, and fibronectin in saltwater drowning and freshwater drowning.
International journal of legal medicine (2019-06-22)
Rosario Barranco, Claudia Castiglioni, Francesco Ventura, Tony Fracasso
ABSTRACT
The diagnosis of drowning is one of the most difficult in forensic medicine. The aim of this study was to analyze pulmonary tissue reactions in death by drowning. In particular, we focused on the immunohistochemical expression of P-selectin, SP-A, HSP70, AQP-5, and fibronectin to investigate our expression in drowning and to understand whether there are differences between saltwater drowning (SWD) and freshwater drowning (FWD), which may indicate a different pathophysiology. We retrospectively investigated 10 cases of SWD (Mediterranean Sea) from the Institute of Legal Medicine of Genoa (Italy), and 10 cases of FWD (Lake of Geneva) from the University Center of Legal Medicine of Geneva (Switzerland). As control group, we examined 10 cases of death by acute external bleeding, characterized by minimal respiratory distress. As compared with controls, in SWD cases, the results showed a decrease of SP-A expression with membrane patterns. Furthermore, we observed a greater SP-A expression with granular pattern in drowning cases without statistically significant difference between SWD and FWD. For the markers AQP-5, HSP70, fibronectin, and P-selectin, no statistically significant differences were found between SWD, FWD, and controls.
Sigma-Aldrich
Anti-Surfactant Protein A Antibody, from rabbit
Sigma-Aldrich
Anti-Hsp70 Mouse mAb (C92F3A-5), liquid, clone C92F3A-5, Calbiochem®