Merck
  • Acute responses of bone specific and related markers to maximal eccentric exercise of the knee extensors and flexors in young men.
Acute responses of bone specific and related markers to maximal eccentric exercise of the knee extensors and flexors in young men.
Journal of musculoskeletal & neuronal interactions (2020-06-03)
Tsang-Hai Huang, Jung-Charng Lin, Mi-Chia Ma, Tsung Yu, Trevor C Chen
ABSTRACT
The present study determined time-course changes in plasma bone-specific and -related markers following a bout of maximal eccentric contractions (MaxEC) of bilateral knee extensors (KE) and flexors (KF). Sedentary young men (n=30) performed a bout of 10 sets of 10 MaxEC (30°/s) of KE and KF with each leg, respectively. Maximal voluntary isometric contraction (MVC) torque, muscle soreness (SOR), plasma creatine kinase (CK) activity, insulin, leptin, tumor necrosis factor-α (TNF-α), undercarboxylated-osteocalcin (ucOCN), carboxy-terminal crosslinking telopeptide of type I collagen (CTX-1) and procollagen type I N-terminal propeptide (P1NP) concentrations were measured from before to 7 days after MaxEC. Significant changes in MVC (KE: -28%, KF: -38%), SOR and plasma CK activity (peak: 39,163 IU/L) following MaxEC were evident (P<0.05) compared to baseline. Plasma leptin (17%) concentrations decreased at 1 day after MaxEC. In bone related markers, plasma ucOCN concentrations (20%) increased at 7 days after MaxEC, and plasma CTX-1 concentrations decreased at 2, 4 and 7 days after MaxEC (6~7%; P<0.05). These results demonstrate that a lean effect of bone generation and an enhanced energy anabolism can be induced by a single bout of MaxEC.