Merck
  • Pre-hybridisation: an efficient way of suppressing endogenous biotin-binding activity inherent to biotin-streptavidin detection system.
Pre-hybridisation: an efficient way of suppressing endogenous biotin-binding activity inherent to biotin-streptavidin detection system.
Journal of immunological methods (2014-03-25)
Raju Ahmed, Emma Spikings, Shaobo Zhou, Andrew Thompsett, Tiantian Zhang
ABSTRACT
Endogenous biotin or biotinylated protein binding activity is a major drawback to biotin-avidin/streptavidin detection system. The avidin/streptavidin conjugate used to detect the complex of the biotinylated secondary antibody and the primary antibody binds to endogenous biotin or biotinylated proteins leading to non-specific signals. In Western blot, the endogenous biotin or biotinylated protein binding activity is usually manifested in the form of ~72kDa, ~75kDa and ~150kDa protein bands, which often mask the signals of interest. To overcome this problem, a method based on prior hybridisation of the biotinylated secondary antibody and the streptavidin conjugate was developed. The method was tested alongside the conventional biotin-streptavidin method on proteins extracted from zebrafish (Danio rerio) embryos. Results showed that the newly developed method efficiently suppresses the endogenous biotin or biotinylated protein binding activity inherent to the biotin-streptavidin detection system.
Sigma-Aldrich
Anti-Pyruvate Carboxylase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.