Merck
  • Anti-inflammatory effect of galectin-1 in a murine model of atopic dermatitis.
Anti-inflammatory effect of galectin-1 in a murine model of atopic dermatitis.
Journal of molecular medicine (Berlin, Germany) (2017-07-01)
Mab Pereira Corrêa, Frans Eberth Costa Andrade, Alexandre Dantas Gimenes, Cristiane Damas Gil
ABSTRACT
Atopic dermatitis (AD) is caused by both dysregulated immune responses and an impaired skin barrier. Although beta-galactoside-binding protein galectin-1 (Gal-1) has immunomodulatory effects in several inflammatory disorders, therapeutic strategies based on its anti-inflammatory properties have not been explored in AD. Thus, we evaluate pharmacological treatment with Gal-1 in the progression of an ovalbumin (OVA)-induced AD-like skin lesions. The skin of OVA-immunized male BALB/c mice was challenged with drops containing OVA on days 11, 14-18 and 21-24. Additionally, in the last week, a subset of animals was treated intraperitoneally with recombinant Gal-1 (rGal-1) or dexamethasone (Dex). Treatment with rGal-1 decreased the clinical signs of dermatitis in BALB/c mice and diminished local eotaxin and IFN-γ levels. The treatment also suppressed the infiltration of eosinophils and mast cells, which was verified by reduced expression of mouse mast cell protease 6 (mMCP6) and eosinophil peroxidase (EPX). These localized effects are associated with extracellular signal-regulated kinase (ERK) activation and downregulation of endogenous Gal-1. The inhibition of disease progression induced by rGal-1 was also correlated with reduced plasma IL-17 levels. Our results demonstrate that rGal-1 is an effective treatment for allergic skin inflammation in AD and may impact the development of novel strategies for skin inflammatory diseases. Pharmacological treatment with rGal-1 reduces clinical signs of atopic dermatitis. Systemic treatment with rGal-1 inhibits eosinophil and mast cell influx in the skin of AD animals. rGal-1 reduced local eotaxin levels and systemic IL-17 levels. The inhibition of disease progression induced by rGal-1 was correlated with upregulation of phosphorylated ERK.