Merck
  • An Investigation into the Immunomodulatory Activities of Sutherlandia frutescens in Healthy Mice.
An Investigation into the Immunomodulatory Activities of Sutherlandia frutescens in Healthy Mice.
PloS one (2016-08-31)
Wei Lei, Jimmy D Browning, Peggy A Eichen, William R Folk, Grace Y Sun, Dennis B Lubahn, Kevin L Fritsche
ABSTRACT
Sutherlandia frutescens is a medicinal plant that has been traditionally used in southern Africa for cancers, infections, and inflammatory conditions. We recently published experiments demonstrating that an aqueous extract of S. frutescens possessed potent immune-stimulatory activity. This work was carried out with murine macrophages, an immune cell type that plays a pivotal role in host defense from infection and in shaping host inflammatory and immune responses. Here, we conducted a series of follow-up experiments to explore the impact of consuming S. frutescens on host response to bacterial challenge using healthy mice. We found that feeding mice a diet containing S. frutescens failed to significantly alter host response to systemic infection by either a gram-positive or gram-negative bacterium (i.e., L. monocytogenes and E. coli, respectively). In contrast to the in vitro observations, we found no evidence that S. frutescens consumption stimulated in vivo inflammatory responses; instead, consumption of S. frutescens tended to diminish in vivo inflammatory responses. Several possible reasons for this are discussed.