Merck
  • Investigation of the role of typhoid toxin in acute typhoid fever in a human challenge model.
Investigation of the role of typhoid toxin in acute typhoid fever in a human challenge model.
Nature medicine (2019-07-05)
Malick M Gibani, Elizabeth Jones, Amber Barton, Celina Jin, Juliette Meek, Susana Camara, Ushma Galal, Eva Heinz, Yael Rosenberg-Hasson, Gerlinde Obermoser, Claire Jones, Danielle Campbell, Charlotte Black, Helena Thomaides-Brears, Christopher Darlow, Christina Dold, Laura Silva-Reyes, Luke Blackwell, Maria Lara-Tejero, Xuyao Jiao, Gabrielle Stack, Christoph J Blohmke, Jennifer Hill, Brian Angus, Gordon Dougan, Jorge Galán, Andrew J Pollard
ABSTRACT
Salmonella Typhi is a human host-restricted pathogen that is responsible for typhoid fever in approximately 10.9 million people annually1. The typhoid toxin is postulated to have a central role in disease pathogenesis, the establishment of chronic infection and human host restriction2-6. However, its precise role in typhoid disease in humans is not fully defined. We studied the role of typhoid toxin in acute infection using a randomized, double-blind S. Typhi human challenge model7. Forty healthy volunteers were randomized (1:1) to oral challenge with 104 colony-forming units of wild-type or an isogenic typhoid toxin deletion mutant (TN) of S. Typhi. We observed no significant difference in the rate of typhoid infection (fever ≥38 °C for ≥12 h and/or S. Typhi bacteremia) between participants challenged with wild-type or TN S. Typhi (15 out of 21 (71%) versus 15 out of 19 (79%); P = 0.58). The duration of bacteremia was significantly longer in participants challenged with the TN strain compared with wild-type (47.6 hours (28.9-97.0) versus 30.3(3.6-49.4); P ≤ 0.001). The clinical syndrome was otherwise indistinguishable between wild-type and TN groups. These data suggest that the typhoid toxin is not required for infection and the development of early typhoid fever symptoms within the context of a human challenge model. Further clinical data are required to assess the role of typhoid toxin in severe disease or the establishment of bacterial carriage.
Sigma-Aldrich
Goat Anti-Human IgM, μ-Chain Specific Alkaline Phosphatase Conjugate, liquid, Calbiochem®
Sigma-Aldrich
Goat Anti-Human IgG, γ-Chain Specific Alkaline Phosphatase Conjugate, liquid, Calbiochem®
Sigma-Aldrich
Goat Anti-Human IgA, α-Chain Specific Alkaline Phosphatase Conjugate, liquid, Calbiochem®