Merck
  • Activation of AMPK suppresses S1P-induced airway smooth muscle cells proliferation and its potential mechanisms.
Activation of AMPK suppresses S1P-induced airway smooth muscle cells proliferation and its potential mechanisms.
Molecular immunology (2020-10-31)
Yilin Pan, Lu Liu, Qianqian Zhang, Wenhua Shi, Wei Feng, Jian Wang, Qingting Wang, Shaojun Li, Manxiang Li
ABSTRACT
The aims of the present study were to investigate the signaling mechanisms for sphingosine-1-phosphate (S1P)-induced airway smooth muscle cells (ASMCs) proliferation and to explore the effect of activation of adenosine monophosphate-activated protein kinase (AMPK) on S1P-induced ASMCs proliferation and its underlying mechanisms. S1P phosphorylated signal transducer and activator of transcription 3 (STAT3) through binding to S1PR2/3, and this further sequentially up-regulated polo-like kinase 1 (PLK1) and inhibitor of differentiation 2 (ID2) protein expression. Pretreatment of cells with S1PR2 antagonist JTE-013, S1PR3 antagonist CAY-10444, knockdown of STAT3, PLK1 and ID2 attenuated S1P-triggered ASMCs proliferation. In addition, activation of AMPK by metformin inhibited S1P-induced ASMCs proliferation by suppressing STAT3 phosphorylation and therefore suppression of PLK1 and ID2 protein expression. Our study suggests that S1P promotes ASMCs proliferation by stimulating S1PR2/3/STAT3/PLK1/ID2 axis, and activation of AMPK suppresses ASMCs proliferation by targeting on STAT3 signaling pathway. Activation of AMPK might benefit asthma by inhibiting airway remodeling.
Sigma-Aldrich
MISSION® esiRNA, targeting human ID2
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human PRKAA2
Pricing and availability is not currently available.