Merck
  • Optimized piperine-phospholipid complex with enhanced bioavailability and hepatoprotective activity.
Optimized piperine-phospholipid complex with enhanced bioavailability and hepatoprotective activity.
Pharmaceutical development and technology (2020-10-17)
Sayan Biswas, Pulok K Mukherjee, Amit Kar, Subhadip Bannerjee, Rawiwan Charoensub, Thidarat Duangyod
ABSTRACT
Piper species is one of the most widely consumed spices for culinary purposes. Piperine (PIP) present in Piper species has a wide range of therapeutic activity including hepatoprotection. However, the major biological limitation of PIP is its low bioavailability after oral administration. Purpose of the study was to prepare an optimized and adequately characterized PIP-phospholipid complex (PPC) as a delivery system to overcome these limitations and to investigate the pharmacokinetics and hepato-protectivity of the formulation in the animal model. Response surface methodology was adopted to optimize the process parameters for PPC preparation. FT-IR, DTA, PXRD, SEM, molecular docking etc. were used for characterization. Solubility, log P, dissolution efficiency and in vivo pharmacokinetics were also investigated. PPC showed enhanced hepatoprotective potential as compared to pure PIP at the same dose level (25 and 50 mg/kg). PPC restored the levels of serum marker and antioxidant enzymes. PPC also increased the bioavailability of PIP in rat serum by 10.40-fold in comparison with pure PIP at the same dose level and enhanced the elimination half-life (t1/2 el) from 0.477 ± 1.76 to 9.80 ± 1.98 h. Results concluded that PPC enhanced the hepatoprotection of PIP which may be due to the improved bioavailability and pharmacokinetics of PIP in rats.
Sigma-Aldrich
Berberine chloride hydrate, technical, ≥90% (AT)
Pricing and availability is not currently available.