Merck
  • Structure-activity relationship studies of flavonoids as potent inhibitors of human platelet 12-hLO, reticulocyte 15-hLO-1, and prostate epithelial 15-hLO-2.
Structure-activity relationship studies of flavonoids as potent inhibitors of human platelet 12-hLO, reticulocyte 15-hLO-1, and prostate epithelial 15-hLO-2.
Bioorganic & medicinal chemistry (2007-09-18)
Yesseny Vasquez-Martinez, Rachana V Ohri, Victor Kenyon, Theodore R Holman, Silvia Sepúlveda-Boza
ABSTRACT
Human lipoxygenase (hLO) isozymes have been implicated in a number of disease states and have attracted much attention with respect to their inhibition. One class of inhibitors, the flavonoids, have been shown to be potent lipoxygenase inhibitors but their study has been restricted to those compounds found in nature, which have limited structural variability. We have therefore carried out a comprehensive study to determine the structural requirements for flavonoid potency and selectivity against platelet 12-hLO, reticulocyte 15-hLO-1, and prostate epithelial 15-hLO-2. We conclude from this study that catechols are essential for high potency, that isoflavones and isoflavonones tend to select against 12-hLO, that isoflavons tend to select against 15-hLO-1, but few flavonoids target 15-hLO-2.
Sigma-Aldrich
Baicalein, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Nordihydroguaiaretic acid, ≥97.0% (HPLC)
Pricing and availability is not currently available.