Merck
  • Identification of benzodiazepine binding site residues in the gamma2 subunit of the gamma-aminobutyric acid(A) receptor.
Identification of benzodiazepine binding site residues in the gamma2 subunit of the gamma-aminobutyric acid(A) receptor.
Molecular pharmacology (2000-04-25)
A M Kucken, D A Wagner, P R Ward, J A Teissére, A J Boileau, C Czajkowski
ABSTRACT
gamma-Aminobutyric acid(A) receptor gamma-subunits are important for benzodiazepine (BZD) binding and modulation of the gamma-aminobutyric acid-mediated Cl(-) current. Previously, by using gamma2/alpha1 chimeric subunits, we identified two domains of the gamma2-subunit, Lys-41-Trp-82 and Arg-114-Asp-161, that are, in conjunction, necessary and sufficient for high-affinity BZD binding. In this study, we generated additional gamma2/alpha1 chimeric subunits and gamma2 point mutants to identify specific residues within the gamma2 Lys-41-Trp-82 region that contribute to BZD binding. Mutant gamma2 and gamma2/alpha1 chimeric subunits were expressed with wild-type alpha1 and beta2 subunits in HEK 293 cells, and the binding of several BZDs was measured. We present evidence that the gamma2 region Met-57-Ile-62 is important for flunitrazepam binding and that, in particular, gamma2 Met-57 and gamma2 Tyr-58 are essential determinants for conferring high-affinity binding. Furthermore, we identify an additional residue, gamma2 Ala-79, that not only is important for high-affinity binding by flunitrazepam (a strong positive modulator) but also plays a crucial role in the binding of the imidazobenzodiazepines Ro15-1788 (a zero modulator) and Ro15-4513 (a weak negative modulator) in the BZD binding pocket. Results from site-directed mutagenesis of gamma2 Ala-79 suggest that this residue may be part of a microdomain within the BZD binding site that is important for binding imidazobenzodiazepines. This separation of drug-specific microdomains for competitive BZD ligands lends insight into the structural determinants governing the divergent effects of these compounds.
Sigma-Aldrich
Ro 15-4513, solid
Pricing and availability is not currently available.