Merck
  • Rapid Quantification of Fungicide Effectiveness on Inhibiting Wheat Stripe Rust Pathogen (Puccinia striiformis f. sp. tritici).
Rapid Quantification of Fungicide Effectiveness on Inhibiting Wheat Stripe Rust Pathogen (Puccinia striiformis f. sp. tritici).
Plant disease (2020-07-11)
Furong Peng, Miu Si, Yang Zizhu, Yuhang Fu, Yuheng Yang, Yang Yu, Chaowei Bi
ABSTRACT
Wheat stripe rust, caused by Puccinia striiformis f. sp. tritici (Pst), is one of the most important and devastating diseases of wheat; therefore, it is necessary to rapidly and accurately quantify fungicide effectiveness to monitor Pst sensitivity and manage the disease. In this study, a rapid method of quantifying the fungicide effectiveness with detached leaves was developed. The results showed that 0.5% water agar containing 75 μg/ml of 6-benzylaminopurine and filter paper worked the best for maintaining wheat leaves. The disease incidences of different concentrations of spore suspension were compared. When the spore concentrations were 5 and 10 mg/ml, the disease incidences had no significant differences at 12 and 15 days after inoculation (P < 0.05). Fungicide treatment tests revealed that there were no significant differences in the efficacies of triadimefon on rust suppression between detached leaves in the culture dishes and direct spray on seedlings. We also developed a Photoshop software method that can replace the current classification method and accurately measure the proportion of sporulation area on infected leaves. The sensitivity baseline of Pst to triadimefon was estimated as 0.1453 ± 0.0081 μg/ml, and all the values of EC50 were tested for normal distribution using the Shapiro-Wilk test (W = 0.204). The baseline can be used to test the sensitivity of different Pst isolates to triadimefon.
Sigma-Aldrich
Gibberellin, 80% gibberellin A3 basis (TLC)
Pricing and availability is not currently available.