Merck
  • Platinum(II) Complexes with Bulky Disubstitute Triazolopyrimidines as Promising Materials for Anticancer Agents.
Platinum(II) Complexes with Bulky Disubstitute Triazolopyrimidines as Promising Materials for Anticancer Agents.
Materials (Basel, Switzerland) (2020-12-02)
Iwona Łakomska, Dariusz Śmiłowicz, Mateusz Jakubowski, Jerzy Sitkowski, Andrzej Wojtczak
ABSTRACT
Herein, we present dicarboxylate platinum(II) complexes of the general formula [Pt(mal)(DMSO)(L)] and [Pt(CBDC)(DMSO)(L)], where L is dbtp 5,7-ditertbutyl-1,2,4-triazolo[1,5-a]pyrimidine) or ibmtp (7-isobutyl-5-methyl-1,2,4- triazolo[1,5-a]pyrimidine), as prospective prodrugs. The platinum(II) complexes were synthesized in a one-pot reaction between cis-[PtCl2(DMSO)2], silver malonate or silver cyclobutane-1,1-dicarboxylate and triazolopyrimidines. All platinum(II) compounds were characterized by FT-IR, and 1H, 13C, 15N and 195Pt NMR; and their square planar geometries with one monodentate N(3)-bonded 5,7-disubstituted-1,2,4-triazolo[1,5-a]pyrimidine, one S-bonded molecule of dimethyl sulfoxide and one O,O-chelating malonato (1, 2) or O,O-chelating cyclobutane-1,1-dicarboxylato (3, 4) was determined. Additionally, [Pt(CBDC)(dbtp)(DMSO)] (3) exhibited (i) substantial in vitro cytotoxicity against the lung adenocarcinoma epithelial cell line (A549) (IC50 = 5.00 µM) and the cisplatin-resistant human ductal breast epithelial tumor cell line (T47D) (IC50 = 6.60 µM); and (ii) definitely exhibited low toxicity against normal murine embryonic fibroblast cells (BALB/3T3).
Sigma-Aldrich
6-Methyl-2,4-heptanedione, ≥98%