Merck
  • Andrographolide and Its 14-Aryloxy Analogues Inhibit Zika and Dengue Virus Infection.
Andrographolide and Its 14-Aryloxy Analogues Inhibit Zika and Dengue Virus Infection.
Molecules (Basel, Switzerland) (2020-11-05)
Feng Li, Wipaporn Khanom, Xia Sun, Atchara Paemanee, Sittiruk Roytrakul, Decai Wang, Duncan R Smith, Guo-Chun Zhou
ABSTRACT
Andrographolide is a labdene diterpenoid with potential applications against a number of viruses, including the mosquito-transmitted dengue virus (DENV). In this study, we evaluated the anti-viral activity of three 14-aryloxy analogues (ZAD-1 to ZAD-3) of andrographolide against Zika virus (ZIKV) and DENV. Interestingly, one analogue, ZAD-1, showed better activity against both ZIKV and DENV than the parental andrographolide. A two-dimension (2D) proteomic analysis of human A549 cells treated with ZAD-1 compared to cells treated with andrographolide identified four differentially expressed proteins (heat shock 70 kDa protein 1 (HSPA1A), phosphoglycerate kinase 1 (PGK1), transketolase (TKT) and GTP-binding nuclear protein Ran (Ran)). Western blot analysis confirmed that ZAD-1 treatment downregulated expression of HSPA1A and upregulated expression of PGK1 as compared to andrographolide treatment. These results suggest that 14-aryloxy analogues of andrographolide have the potential for further development as anti-DENV and anti-ZIKV agents.
Sigma-Aldrich
Anti-HSPA1A antibody produced in rabbit, IgG fraction of antiserum
Pricing and availability is not currently available.