Merck
  • Designed CXCR4 mimic acts as a soluble chemokine receptor that blocks atherogenic inflammation by agonist-specific targeting.
Designed CXCR4 mimic acts as a soluble chemokine receptor that blocks atherogenic inflammation by agonist-specific targeting.
Nature communications (2020-11-27)
Christos Kontos, Omar El Bounkari, Christine Krammer, Dzmitry Sinitski, Kathleen Hille, Chunfang Zan, Guangyao Yan, Sijia Wang, Ying Gao, Markus Brandhofer, Remco T A Megens, Adrian Hoffmann, Jessica Pauli, Yaw Asare, Simona Gerra, Priscila Bourilhon, Lin Leng, Hans-Henning Eckstein, Wolfgang E Kempf, Jaroslav Pelisek, Ozgun Gokce, Lars Maegdefessel, Richard Bucala, Martin Dichgans, Christian Weber, Aphrodite Kapurniotu, Jürgen Bernhagen
ABSTRACT
Targeting a specific chemokine/receptor axis in atherosclerosis remains challenging. Soluble receptor-based strategies are not established for chemokine receptors due to their discontinuous architecture. Macrophage migration-inhibitory factor (MIF) is an atypical chemokine that promotes atherosclerosis through CXC-motif chemokine receptor-4 (CXCR4). However, CXCR4/CXCL12 interactions also mediate atheroprotection. Here, we show that constrained 31-residue-peptides ('msR4Ms') designed to mimic the CXCR4-binding site to MIF, selectively bind MIF with nanomolar affinity and block MIF/CXCR4 without affecting CXCL12/CXCR4. We identify msR4M-L1, which blocks MIF- but not CXCL12-elicited CXCR4 vascular cell activities. Its potency compares well with established MIF inhibitors, whereas msR4M-L1 does not interfere with cardioprotective MIF/CD74 signaling. In vivo-administered msR4M-L1 enriches in atherosclerotic plaques, blocks arterial leukocyte adhesion, and inhibits atherosclerosis and inflammation in hyperlipidemic Apoe-/- mice in vivo. Finally, msR4M-L1 binds to MIF in plaques from human carotid-endarterectomy specimens. Together, we establish an engineered GPCR-ectodomain-based mimicry principle that differentiates between disease-exacerbating and -protective pathways and chemokine-selectively interferes with atherosclerosis.
Sigma-Aldrich
Fmoc-6-Ahx-OH, ≥97.0% (HPLC)
Pricing and availability is not currently available.