Merck
  • Comparison of CD63 Upregulation Induced by NSAIDs on Basophils and Monocytes in Patients with NSAID Hypersensitivity.
Comparison of CD63 Upregulation Induced by NSAIDs on Basophils and Monocytes in Patients with NSAID Hypersensitivity.
Journal of allergy (2011-12-22)
N Abuaf, H Rostane, J Barbara, C Toly-Ndour, H Gaouar, P Mathelier-Fusade, F Leynadier, C Francès, R Girot
ABSTRACT
Background. An in vitro basophil activation test, based on the detection of CD63 upregulation induced by NSAIDs, has been described. Its clinical significance remains controversial. Objectives. In patients with a history of nonallergic NSAID hypersensitivity, stratified according to the severity of the symptoms, to assess with NSAIDs the predictive value of basophil (BAT) and monocyte (MAT) activation tests. Patients/Methods. Sixty patients who had NSAIDs-induced or exacerbated urticaria/angiooedema and 20 controls was included. After incubation with NSAIDs or acetaminophen, leukocytes were analysed for CD63 upregulation. Results. With aspirin, the sensitivity (37%) and specificity (90%) of BAT agree with already published results. In contrast, when patients had had cutaneous and visceral reactions, the frequency of positive BAT 14/22 (64%, P < 0.001) or MAT 10/22 (46%, P < 0.01) were increased. Conclusions. Positive tests were more frequent among patients having a severe hypersensitivity contrasting with the other patients who had results similar to controls.
Product Description
Supelco
Calcium ionophore V, Selectophore, ≥95%
Supelco
Calcium ionophore I, Selectophore, for ion-selective electrodes, function tested, ≥99.0%