Merck
  • Inhibitory effect of noradrenaline uptake inhibitors on contractions of rat aortic smooth muscle.
Inhibitory effect of noradrenaline uptake inhibitors on contractions of rat aortic smooth muscle.
British journal of pharmacology (1996-02-01)
Y Huang
ABSTRACT
1 The effects of noradrenaline (NA) uptake inhibitors on contractions induced by NA, high K+, and 12-O-tetradecanoylphorbol-13-acetate (TPA) in rat isolated aorta were investigated. 2 Protriptyline (0.3 microM) and amitriptyline (0.3 microM) produced an approximately parallel shift to the right in the dose-response curve to NA. Protriptyline (> 0.3 microM), amitriptyline (> 0.3 microM) and xylamine (0.01-1 microM) significantly reduced the maximal contractile response to NA. The IC50 values for inhibition of the contractile response to 3 microM NA were 1.58 microM for xylamine, 1.70 microM for amitriptyline and 2.57 microM for protriptyline. 3 Protriptyline and amitriptyline dose-dependently inhibited the high K+ (60 mM)-induced contraction (IC50 = 0.69 microM for protriptyline and IC50 = 3.15 microM for amitriptyline). In contrast, xylamine did not affect the high K(+)-induced contraction. 4 Protriptyline and amitriptyline dose-dependently inhibited TPA (1 microM)-induced contraction in calcium-free solution; xylamine (up to 30 microM) was without effect. Staurosporine (10 nM) completely inhibited the TPA- and NA-induced contraction. 5. Protriptyline (3 microM) and amitriptyline (3 microM) caused about 54% and 60% inhibition, respectively, of aortic contractions caused by endothelin-1 (10 nM) in the absence of endothelium. Xylamine (10 microM) was without effect. 6 Inhibitory effects of NA uptake inhibitors on contractions were independent of the presence of endothelium and were unaffected by the K+ channel blockers, tetraethylammonium ions (up to 3 mM) and glibenclamide (up to 30 microM). 7 These results indicate that tricyclic antidepressant drugs such as protriptyline and amitriptyline could act as both postsynaptic adrenoceptor antagonists and direct inhibitors of muscle contraction; whereas, xylamine, a structurally distinct NA uptake blocker might principally exert its action only at alpha-adrenoceptors on rat aortic smooth muscle.
Product Description
Supelco
Protriptyline hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.