Merck
  • Gold Nanorod-based Photo-PCR System for One-Step, Rapid Detection of Bacteria.
Gold Nanorod-based Photo-PCR System for One-Step, Rapid Detection of Bacteria.
Nanotheranostics (2017-10-27)
Jinjoo Kim, Hansol Kim, Ji Ho Park, Sangyong Jon
ABSTRACT
The polymerase chain reaction (PCR) has been an essential tool for diagnosis of infectious diseases, but conventional PCR still has some limitations with respect to applications to point-of-care (POC) diagnostic systems that require rapid detection and miniaturization. Here we report a light-based PCR method, termed as photo-PCR, which enables rapid detection of bacteria in a single step. In the photo-PCR system, poly(enthylene glycol)-modified gold nanorods (PEG-GNRs), used as a heat generator, are added into the PCR mixture, which is subsequently periodically irradiated with a 808-nm laser to create thermal cycling. Photo-PCR was able to significantly reduce overall thermal cycling time by integrating bacterial cell lysis and DNA amplification into a single step. Furthermore, when combined with KAPA2G fast polymerase and cooling system, the entire process of bacterial genomic DNA extraction and amplification was further shortened, highlighting the potential of photo-PCR for use in a portable, POC diagnostic system.
Product Description
Roche
KAPA2G Fast PCR Kit
Pricing and availability is not currently available.