Merck
  • Comparative study between topically applied irradiated human amniotic membrane in combination with tea tree oil versus topical tioconazole in pityraisis versicolor treatment.
Comparative study between topically applied irradiated human amniotic membrane in combination with tea tree oil versus topical tioconazole in pityraisis versicolor treatment.
Cell and tissue banking (2020-03-13)
Radwan K Nashwa, El Bedewi Ahmed, Waleed A Nemr
ABSTRACT
Pityriasis versicolor (PV) is a chronic skin disease caused by virulence activities of Malassezia, a genus of skin-associated yeasts. Traditionally, Tioconazole is used as a topical antifungal for curing PV. Previous investigations cited that human amniotic membrane (HAM), a placental tissue, has antimicrobial and anti-inflammatory activities and is useful as a dressing for healing skin lesions. Moreover, tea tree oil (TTO) has a potent antifungal efficacy. This clinical trial aims to achieve an alternative therapeutic treatment able to kill Malassezia and heal PV lesions using TTO-saturated HAM (TOSHAM), with little application times. This study subjected 120 patients with hypopigmented or hyperpigmented PV lesions; half patients were treated weekly with TOSHAM compared with the others who applying 1% Tioconazole cream daily as a traditional treatment. Microbiological evaluation of in vitro fungicidal activity of TOSHAM versus Tioconazole was carried out against Malassezia furfur culture. The clinical outcomes of this study proved the superior activity of TOSHAM to heal PV lesions than Tioconazole; this was in harmony with microbiological findings. This study approached a novel therapeutic treatment of PV with great outcomes by using TOSHAM.
Sigma-Aldrich
Minimum Essential Medium Eagle, With Hanks′ salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Pricing and availability is not currently available.