Merck
  • Discovery of novel West Nile Virus protease inhibitor based on isobenzonafuranone and triazolic derivatives of eugenol and indan-1,3-dione scaffolds.
Discovery of novel West Nile Virus protease inhibitor based on isobenzonafuranone and triazolic derivatives of eugenol and indan-1,3-dione scaffolds.
PloS one (2019-09-27)
André S de Oliveira, Poliana A R Gazolla, Ana Flávia C da S Oliveira, Wagner L Pereira, Lívia C de S Viol, Angélica F da S Maia, Edjon G Santos, Ítalo E P da Silva, Tiago A de Oliveira Mendes, Adalberto M da Silva, Roberto S Dias, Cynthia C da Silva, Marcelo D Polêto, Róbson R Teixeira, Sergio O de Paula
ABSTRACT
The West Nile Virus (WNV) NS2B-NS3 protease is an attractive target for the development of therapeutics against this arboviral pathogen. In the present investigation, the screening of a small library of fifty-eight synthetic compounds against the NS2-NB3 protease of WNV is described. The following groups of compounds were evaluated: 3-(2-aryl-2-oxoethyl)isobenzofuran-1(3H)-ones; eugenol derivatives bearing 1,2,3-triazolic functionalities; and indan-1,3-diones with 1,2,3-triazolic functionalities. The most promising of these was a eugenol derivative, namely 4-(3-(4-allyl-2-methoxyphenoxy)-propyl)-1-(2-bromobenzyl)-1H-1,2,3-triazole (35), which inhibited the protease with IC50 of 6.86 μmol L-1. Enzyme kinetic assays showed that this derivative of eugenol presents competitive inhibition behaviour. Molecular docking calculations predicted a recognition pattern involving the residues His51 and Ser135, which are members of the catalytic triad of the WNV NS2B-NS3 protease.
Sigma-Aldrich
4-Pentyn-1-ol, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
1,3-Indandione, 97%
Pricing and availability is not currently available.