Merck
  • Synovial fluid lubricin and hyaluronan are altered in equine osteochondral fragmentation, cartilage impact injury, and full-thickness cartilage defect models.
Synovial fluid lubricin and hyaluronan are altered in equine osteochondral fragmentation, cartilage impact injury, and full-thickness cartilage defect models.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society (2020-01-23)
Bridgette T Peal, Rachel Gagliardi, Jin Su, Lisa A Fortier, Michelle L Delco, Alan J Nixon, Heidi L Reesink
ABSTRACT
The objectives of this study were to evaluate temporal changes in lubricin, hyaluronan (HA), and HA molecular weight (MW) distributions in three distinct models of equine joint injury affecting the carpal (wrist), tarsal (ankle), and femoropatellar (knee) joints. To establish ranges for lubricin, HA, and HA MW distributions across multiple joints, we first evaluated clinically healthy, high-motion equine joints. Synovial fluid was collected from high-motion joints in horses without clinical signs of joint disease (n = 11 horses, 102 joints) and from research horses undergoing carpal osteochondral fragmentation (n = 8), talar cartilage impact injury (n = 7), and femoral trochlear ridge full-thickness cartilage injury (n = 22) prior to and following arthroscopically induced joint injury. Lubricin and HA concentrations were measured via enzyme-linked immunosorbent assays, and gel electrophoresis was performed to evaluate HA MW distributions. Synovial fluid parameters were analyzed via linear regression models, revealing that lubricin and HA concentrations were conserved across healthy, high-motion joints. Lubricin concentrations increased post-injury in all osteoarthritis models (carpal fragmentation P = .001; talar impact P < .001; femoral trochlear ridge cartilage defect P = .03). Sustained loss of HA was noted post-arthroscopy following carpal osteochondral fragmentation (P < .0001) and talar impact injury (P < .001). Lubricin may be elevated to compensate for the loss of HA and to protect cartilage post-injury. Further investigation into the mechanisms regulating lubricin and HA following joint injury and their effects on joint homeostasis is warranted, including whether lubricin has value as a biomarker for post-traumatic osteoarthritis.
Sigma-Aldrich
Anti-Lubricin Antibody/Proteoglycan 4, clone 9G3, clone 9G3, from mouse