Merck
  • Endothelin receptor heteromerization inhibits β-arrestin function in HEK293 cells.
Endothelin receptor heteromerization inhibits β-arrestin function in HEK293 cells.
Canadian journal of physiology and pharmacology (2020-08-04)
Adel Zrein, Amina M Bagher, Alexander P Young, Eileen M Denovan-Wright, Melanie E M Kelly
ABSTRACT
The endothelin receptor A (ETA) and endothelin receptor B (ETB) are G protein-coupled receptors that are co-expressed in vascular smooth muscle cells. Endothelin-1 (ET-1) activates endothelin receptors to cause microvascular vasoconstriction. Previous studies have shown that heteromerization between ETA and ETB prolongs Ca2+ transients, leading to prolongation of Gαq-dependent signaling and sustained vasoconstriction. We hypothesized that these effects are in part mediated by the resistance of ETA/ETB heteromers to β-arrestin recruitment and subsequent desensitization. Using bioluminescence resonance energy transfer 2 (BRET2), we found that ETB has a relatively equal affinity to form either homomers or heteromers with ETA when co-expressed in the human embryonic kidney 293 (HEK293) cells. When co-expressed, activation of ETA and ETB by ET-1 caused a heteromer-specific reduction and delay in β-arrestin-2 recruitment with a corresponding reduction and delay in ET-1-induced ETA/ETB co-internalization. Furthermore, the co-expression of ETA and ETB inhibited ET-1-induced β-arrestin-1-dependent extracellular signal-regulated kinase (ERK) phosphorylation while prolonging ET-1-induced Gαq-dependent ERK phosphorylation. ETA/ETB heteromerization mediates the long-lasting vasoconstrictor response to ET-1 by the prolongation of Gαq-dependent signaling and inhibition of β-arrestin function.
Sigma-Aldrich
Transcreener® AMP2/GMP2 TR-FRET Assay
Sigma-Aldrich
L-(−)-Glucose, ≥99%