Merck
  • Multi-Responsive Polysiloxane/Poly(N-isopropylacrylamide) Interpenetrating Networks Containing Urea and Thiourea Groups.
Multi-Responsive Polysiloxane/Poly(N-isopropylacrylamide) Interpenetrating Networks Containing Urea and Thiourea Groups.
Polymers (2020-05-24)
Junta Sano, Shigeki Habaue
ABSTRACT
Novel interpenetrating polymer networks (IPNs) were synthesized from N-isopropylacrylamide (NIPAM) and polysiloxanes containing a urea or thiourea side group, in addition to the silanol residue, through two reactions, such as the radical gelation of NIPAM and the condensation of silanols to form a siloxane linkage. The obtained IPNs showed a typical temperature-responsive volume change in water based on the constructed poly-NIPAM gel component. In addition, the characteristic color and volume changes responding to chemical stimuli, such as acetate and/or fluoride ions, based on the introduced urea and thiourea groups onto the polysiloxane gel were observed in N,N-dimethylformamide.
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile) solution, 0.2 M in toluene
Sigma-Aldrich
4-Nitrophenyl isothiocyanate, 98%