Merck
  • Investigation of antioxidant effects of rosmarinic acid on liver, lung and kidney in rats: a biochemical and histopathological study.
Investigation of antioxidant effects of rosmarinic acid on liver, lung and kidney in rats: a biochemical and histopathological study.
Folia morphologica (2019-08-23)
A Oğuz, A Böyük, A Ekinci, U Alabalik, A Türkoğlu, M C Tuncer, A Ekingen, E Deveci, B Gültürk, U Aday
ABSTRACT
The aim of the study was to investigate the protective effects of rosmarinic acid in rats exposed to hepatic ischaemia/reperfusion (I/R) injury. Thirty-two rats were randomly classified into four groups of 8 rats each: laparotomy without medication, rosmarinic acid (dose of 50 mg/kg via oral gavage) followed by laparotomy, laparotomy followed by hepatic I/R, and hepatic I/R with rosmarinic acid. Serum aspartate aminotransferase, alaninę aminotransferase, and malondialdehyde levels and total oxidant activity and total antioxidant capacity levels of the liver, lung, and kidney were assessed. The histopathologic assessment was also performed. Rosmarinic acid significantly reduced liver function test parameters and decreased oxidative stress and abnormal histopathologic findings in the liver. The oxidative stress in the lung significantly increased in the I/R group but significantly decreased in the I/R + rosmarinic acid group due to the addition of rosmarinic acid. Rosmarinic acid led to no reduction in oxidative stress in kidney following hepatic I/R injury. There were no statistically significant differences among the groups regarding histopathologic changes in kidney and lung sections. Rosmarinic acid has antioxidant properties and is an effective hepatoprotective agent. However, although rosmarinic acid provides useful effects in the lung by increasing antioxidant capacity and reducing oxidative stress after I/R injury, it does not ameliorate histopathologic changes. These findings suggest that rosmarinic acid is likely to provide favourable outcomes in the treatment of hepatic I/R injury.
Sigma-Aldrich
Rosmarinic acid, ≥98% (HPLC), from Rosemarinus officinalis L.