Merck
  • Guanidinium-Assisted Surface Matrix Engineering for Highly Efficient Perovskite Quantum Dot Photovoltaics.
Guanidinium-Assisted Surface Matrix Engineering for Highly Efficient Perovskite Quantum Dot Photovoltaics.
Advanced materials (Deerfield Beach, Fla.) (2020-05-26)
Xufeng Ling, Jianyu Yuan, Xuliang Zhang, Yuli Qian, Shaik M Zakeeruddin, Bryon W Larson, Qian Zhao, Junwei Shi, Jiacheng Yang, Kang Ji, Yannan Zhang, Yongjie Wang, Chunyang Zhang, Steffen Duhm, Joseph M Luther, Michael Grätzel, Wanli Ma
ABSTRACT
Metal halide perovskite quantum dots (Pe-QDs) are of great interest in new-generation photovoltaics (PVs). However, it remains challenging in the construction of conductive and intact Pe-QD films to maximize their functionality. Herein, a ligand-assisted surface matrix strategy to engineer the surface and packing states of Pe-QD solids is demonstrated by a mild thermal annealing treatment after ligand exchange processing (referred to as "LE-TA") triggered by guanidinium thiocyanate. The "LE-TA" method induces the formation of surface matrix on CsPbI3 QDs, which is dominated by the cationic guanidinium (GA+ ) rather than the SCN- , maintaining the intact cubic structure and facilitating interparticle electrical interaction of QD solids. Consequently, the GA-matrix-confined CsPbI3 QDs exhibit remarkably enhanced charge mobility and carrier diffusion length compared to control ones, leading to a champion power conversion efficiency of 15.21% when assembled in PVs, which is one of the highest among all Pe-QD solar cells. Additionally, the "LE-TA" method shows similar effects when applied to other Pe-QD PV systems like CsPbBr3 and FAPbI3 (FA = formamidinium), indicating its versatility in regulating the surfaces of various Pe-QDs. This work may afford new guidelines to construct electrically conductive and structurally intact Pe-QD solids for efficient optoelectronic devices.
Sigma-Aldrich
Formamidine acetate salt, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Tris(pentafluorophenyl)borane, 95%
Pricing and availability is not currently available.