Merck
  • Modulation of gastrointestinal digestion of β-lactoglobulin and micellar casein following binding by (-)-epigallocatechin-3-gallate (EGCG) and green tea flavanols.
Modulation of gastrointestinal digestion of β-lactoglobulin and micellar casein following binding by (-)-epigallocatechin-3-gallate (EGCG) and green tea flavanols.
Food & function (2020-06-20)
Özge Dönmez, Burçe Ataç Mogol, Vural Gökmen, Ning Tang, Mogens Larsen Andersen, Dereck E W Chatterton
ABSTRACT
The effect of binding of flavonoids, (-)-epigallocatechin-3-gallate (EGCG) and green tea extract (GTE), to beta-lactoglobulin (β-Lg) and micellar casein (micellar casein isolate, MCI) on protein digestibility was investigated. β-Lg resisted digestion by pepsin, but in the presence of EGCG the digestion of β-Lg was enhanced. Binding of EGCG to β-Lg was identified by nitro blue tetrazolium (NBT) staining and found, by isothermal titration calorimetry, to be an enthalpy-driven exothermic process, with a binding constant of 19 950 L mol-1. Binding promoted a more rapid digestion of β-Lg during simulated upper duodenal digestion. NBT staining indicated a loss of binding of EGCG to β-Lg during combined gastric and distal small intestinal digestion and correlated with the cleavage of β-Lg. However, increased β-Lg heteromer formation and reduced β-Lg monomer digestibility were observed for the β-Lg-GTE complex. MCI was more digestible than β-Lg during pepsin digestion, but reduced digestibility was observed for both MCI-EGCG and MCI-GTE complexes, with loss of binding during intestinal digestion. The free radical scavenging capacity (FRSC) of EGCG remained stable for the β-Lg-EGCG complex throughout the gastric and intestinal phases of digestion, but this was significantly lowered for the MCI-EGCG complex. These results indicated that polyphenols bind to milk proteins modulating the in vitro digestibility and FRSC of β-Lg and MCI as a result of the formation of complexes.
Sigma-Aldrich
β-Lactoglobulin from bovine milk, ≥90% (PAGE), lyophilized powder
Pricing and availability is not currently available.
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Pricing and availability is not currently available.