Merck
  • Bone morphogenetic protein 1 cleaves the linker region between ligand-binding repeats 4 and 5 of the LDL receptor and makes the LDL receptor non-functional.
Bone morphogenetic protein 1 cleaves the linker region between ligand-binding repeats 4 and 5 of the LDL receptor and makes the LDL receptor non-functional.
Human molecular genetics (2019-10-11)
Thea Bismo Strøm, Katrine Bjune, Trond P Leren
ABSTRACT
The cell-surface low-density lipoprotein receptor (LDLR) internalizes low-density lipoprotein (LDL) by receptor-mediated endocytosis and plays a key role in the regulation of plasma cholesterol levels. The ligand-binding domain of the LDLR contains seven ligand-binding repeats of approximately 40 residues each. Between ligand-binding repeats 4 and 5, there is a 10-residue linker region that is subject to enzymatic cleavage. The cleaved LDLR is unable to bind LDL. In this study, we have screened a series of enzyme inhibitors in order to identify the enzyme that cleaves the linker region. These studies have identified bone morphogenetic protein 1 (BMP1) as being the cleavage enzyme. This conclusion is based upon the use of the specific BMP1 inhibitor UK 383367, silencing of the BMP1 gene by the use of siRNA or CRISPR/Cas9 technology and overexpression of wild-type BMP1 or the loss-of-function mutant E214A-BMP1. We have also shown that the propeptide of BMP1 has to be cleaved at RSRR120↓ by furin-like proprotein convertases for BMP1 to have an activity towards the LDLR. Targeting BMP1 could represent a novel strategy to increase the number of functioning LDLRs in order to lower plasma LDL cholesterol levels. However, a concern by using BMP1 inhibitors as cholesterol-lowering drugs could be the risk of side effects based on the important role of BMP1 in collagen assembly.
Sigma-Aldrich
MISSION® esiRNA, targeting human BMP1
Sigma-Aldrich
CRISPR/Cas9 Products and Services, Design and order CRISPR gRNA, Cas9, screening libraries, controls and companion products. Formats include plant, lentivirus, IVT-RNA, plasmid, synthetic, and protein.