Merck
  • Safety, Tolerability, Pharmacokinetics, and Pharmacodynamics of the Novel Non-Bile Acid FXR Agonist Tropifexor (LJN452) in Healthy Volunteers.
Safety, Tolerability, Pharmacokinetics, and Pharmacodynamics of the Novel Non-Bile Acid FXR Agonist Tropifexor (LJN452) in Healthy Volunteers.
Clinical pharmacology in drug development (2019-12-12)
Michael K Badman, Jin Chen, Sachin Desai, Soniya Vaidya, Srikanth Neelakantham, Jie Zhang, Lu Gan, Kate Danis, Bryan Laffitte, Lloyd B Klickstein
ABSTRACT
Tropifexor (LJN452) is a potent, orally available, non-bile acid farnesoid X receptor agonist under clinical development for chronic liver diseases. Here, we present results from a first-in-human study of tropifexor following single- and multiple-ascending doses (SAD/MAD) and food effect substudy in healthy volunteers. The SAD study included 6 fasted cohorts receiving 10- to 3000-µg tropifexor or placebo and 1 cohort receiving 300-µg tropifexor with a high-fat meal. The MAD study included 4 lean cohorts receiving 10 to 100 µg and 1 obese cohort receiving 30-µg once-daily doses or placebo for 14 days. Pharmacodynamic assessment of fibroblast growth factor 19 and fasting plasma lipids was performed after dosing. Overall, 95 volunteers received at least 1 tropifexor or placebo dose. Tropifexor was well tolerated up to 3000 µg and 100 µg in the SAD and MAD studies, respectively; however, 2 subjects discontinued the MAD study due to asymptomatic elevation of liver transaminases. At single doses, tropifexor showed a moderate rate of absorption (median time to maximum concentration, 4 hours), dose-proportional increases in exposure, and elimination half-life of 13.5 to 21.9 hours. When taken with food, tropifexor exposure increased by ∼60%. With multiple dosing, steady state was reached on day 4 with <2-fold accumulation. Single and multiple doses showed dose-dependent increases in fibroblast growth factor 19. No changes in serum lipids were observed in tropifexor- vs placebo-treated obese subjects. In conclusion, tropifexor was well tolerated, had a pharmacokinetic profile suitable for once-daily dosing and showed dose-dependent target engagement without altering plasma lipids in healthy volunteers.
Product Description
Supelco
7α-Hydroxy-4-cholesten-3-one, ≥95.0% (HPLC)
Pricing and availability is not currently available.