Merck
  • Disruption of the Notch pathway aggravates airway inflammation by inhibiting regulatory T cell differentiation via regulation of plasmacytoid dendritic cells.
Disruption of the Notch pathway aggravates airway inflammation by inhibiting regulatory T cell differentiation via regulation of plasmacytoid dendritic cells.
Scandinavian journal of immunology (2020-03-19)
Shuo-Yao Qu, Xin-Yu Ti, Jian Zhang, Chang-Gui Wu
ABSTRACT
Plasmacytoid dendritic cells (pDCs) regulate immunity and promote tolerance in asthma. Notch signalling is a highly conserved pathway that regulates the immune response; however, its role in pDC-mediated asthmatic airway inflammation is unclear. This study clarified the effects of Notch signalling on pDC-mediated airway inflammation using murine models of ovalbumin-sensitized allergic asthma. RBP-J-deficient pDCs (RBP-J-/- pDCs) were co-cultured with naïve CD4+ T cells and supernatants and T cell subtypes were analysed. RBP-J-/- pDCs were intranasally transferred to the airways of ovalbumin-sensitized recipient mice. Lung samples of all mice were subjected to tests for histopathology, cytokine profile of bronchoalveolar lavage fluid, airway hyperactivity and expression of T helper type 1 (Th1)/Th2 cells, regulatory T cells and type 2 innate lymphoid cells (ILC2s). The results showed that pDCs with and without RBP-J deficiency significantly differed in expression levels of cluster of differentiation 83 (CD83), but not CD80, CD86 and major histocompatibility complex class II. Co-culturing pDCs with naïve T cells revealed a poorer immunosuppressive effect of RBP-J-/- pDCs. This may be attributed to the lower expression levels of inducible co-stimulator ligand and lower production of interleukin 10 in RBP-J-/- pDCs than in control pDCs, which impeded T cell activation and Treg suppression. RBP-J-/- pDCs were associated with high ILC2 expression and severe Th2 immune responses and airway inflammation. Therefore, Notch signalling is critical for pDC-dependent immunoregulation, and RBP-J deficiency reduces pDC-based immunosuppression via T cell activation and Th cell differentiation. Thus, this pathway may be a therapeutic target for pDC-based anti-asthma immunotherapy.
Sigma-Aldrich
Resiquimod, ≥98% (HPLC)
Sigma-Aldrich
Aluminum hydroxide hydrate