Merck
  • Synovial IL-21/TNF-producing CD4+ T cells induce joint destruction in rheumatoid arthritis by inducing matrix metalloproteinase production by fibroblast-like synoviocytes.
Synovial IL-21/TNF-producing CD4+ T cells induce joint destruction in rheumatoid arthritis by inducing matrix metalloproteinase production by fibroblast-like synoviocytes.
Journal of leukocyte biology (2016-10-14)
Maria C Lebre, Pedro L Vieira, Man Wai Tang, Saïda Aarrass, Boy Helder, Thomas Newsom-Davis, Paul P Tak, Gavin R Screaton
ABSTRACT
Bone and cartilage destruction is one of the key manifestations of rheumatoid arthritis (RA). Although the role of T helper (Th)17 cells in these processes is clear, the role of IL-21-producing cells T cells has been neglected. We sought to investigate the role of IL-21 in RA by focusing on the functional characteristics of the main producers of this cytokine, synovial CD4+IL-21+ T cells. We show that the frequency of both synovial fluid (SF) CD4+IL-21+ or CD4+IL-21+TNF+ T cells in patients with RA was significantly higher compared with patients with psoriatic arthritis (PsA). The frequency of peripheral blood (PB) IL-21+CD4+ T cells in patients with RA positively correlated with disease activity score 28 (DAS28), serum anticyclic citrullinated peptide (anti-CCP) antibodies and IgM-rheumatoid factor (IgM-RF). IL-21 levels in RA SF were associated with matrix metalloproteinase (MMP)-1 and MMP-3. Related to this, IL-21 induced significantly the secretion of MMP-1 and MMP-3 in RA synovial biopsies. Sorted SF CD4+IL-21+ T cells significantly induced the release of MMP-1 and MMP-3 by fibroblast-like synoviocytes (FLS) compared with medium or CD4+IL-21- T cells in a coculture system. Neutralization of both IL-21 and TNF resulted in significantly less production of MMP by FLS. The results of this study indicate a new role for synovial CD4+IL-21+TNF+ T cells in promoting synovial inflammation/joint destruction in patients with RA. Importantly, IL-21 blockade in combination with anti-TNF might be an effective therapy in patients with RA by inhibiting MMP-induced inflammation/joint destruction.
Sigma-Aldrich
Anti-CD28 Antibody, clone 15E8, clone 15E8, Chemicon®, from mouse
Pricing and availability is not currently available.