Merck
  • Anti-allergic effect of the flavonoid myricitrin from Myrica rubra leaf extracts in vitro and in vivo.
Anti-allergic effect of the flavonoid myricitrin from Myrica rubra leaf extracts in vitro and in vivo.
Natural product research (2011-02-18)
Shunsuke Shimosaki, Yoko Tsurunaga, Hiroyuki Itamura, Morihiko Nakamura
ABSTRACT
Flavonoids are ingested by the general population as anti-oxidant and anti-inflammatory agents. In this study, we investigated the effects of myricitrin, a flavonoid rich in Myrica rubra leaf, upon anti-inflammatory action. Myrica rubra leaf extracts inhibited pro-inflammatory TNFα production in a macrophage cell line, Raw264.7 cells. We observed that the serum IgE levels in the leaf extract-treated DO11.10, a mouse allergy model, were down-regulated. HPLC was performed to demonstrate that M. rubra leaf extracts contain a large amount of myricitrin. We observed an inhibitory effect of HPLC-purified myricitrin on TNFα production in Raw264.7 cells. Thus, myricitrin may be of potential interest in the management of inflammatory conditions.
Product Description
Supelco
Myricitrin, analytical standard
Sigma-Aldrich
Myricitrin, ≥99.0% (HPLC)