Merck
  • A simple HPLC method for analysing diaminopimelic acid diastereomers in cell walls of Gram-positive bacteria.
A simple HPLC method for analysing diaminopimelic acid diastereomers in cell walls of Gram-positive bacteria.
Letters in applied microbiology (2000-04-04)
J McKerrow, S Vagg, T McKinney, E M Seviour, A M Maszenan, P Brooks, R J Seviour
ABSTRACT
A simple and sensitive method for separating and detecting the LL, DD and meso diastereomers of the dibasic amino acid diaminopimelic acid (DAP) in the peptidoglycan of Gram-positive bacteria is described. This method is based on reverse phase HPLC separation of chiral derivatives of DAP followed by fluorescence detection of the o-phthaldehyde derivatives. Its application to the analyses of cell walls of several Gram-positive bacteria is described, where 10 mg or less of dry cells is required.
Sigma-Aldrich
DL-2,6-Diaminopimelic acid, ≥95% (TLC)
Sigma-Aldrich
meso-2,6-Diaminopimelic acid, ≥98% (TLC)