Merck
  • Determination of halofantrine and its main metabolite desbutylhalofantrine in rat plasma by high-performance liquid chromatography with on-line UV irradiation and peroxyoxalate chemiluminescence detection.
Determination of halofantrine and its main metabolite desbutylhalofantrine in rat plasma by high-performance liquid chromatography with on-line UV irradiation and peroxyoxalate chemiluminescence detection.
Biomedical chromatography : BMC (2008-10-01)
Abena Amponsaa-Karikari, Naoya Kishikawa, Kaname Ohyama, Kenichiro Nakashima, Naotaka Kuroda
ABSTRACT
A sensitive, selective and reliable method has been developed and validated for the determination of halofantrine and its metabolite desbutylhalofantrine in rat plasma using 9,10-diphenylanthracene as an internal standard. The method is based on peroxyoxalate chemiluminescence detection of hydrogen peroxide produced from fused aromatic rings in the structures of halofantrine and desbutylhalofantrine upon UV irradiation. Using spiked rat plasma, good linear relationships were obtained for both halofantrine and desbutylhalofantrine between peak height ratios (vs internal standard) and their corresponding concentrations over a range of 0.01-0.8 microg/mL with correlation coefficients of at least 0.997. The detection limits at signal-to-noise ratio of 3 using 0.2 mL of rat plasma were 1.5 and 1.4 ng/mL for halofantrine and desbutylhalofantrine, respectively. Relative standard deviations (n = 3) intra- and inter-day were between 0.5 and 5.4% for all the studied concentrations. Using this method with simple sample treatment, halofantrine and desbutylhalofantrine in rat plasma could be precisely determined without interference from endogenous substances. The method was successfully applied to the measurement of the time courses of plasma halofantrine concentration after oral administration of the drug (7 mg/kg) to rats.
Product Description
Supelco
9,10-Diphenylanthracene, analytical standard