Merck
  • HSPA12A Is a Novel Player in Nonalcoholic Steatohepatitis via Promoting Nuclear PKM2-Mediated M1 Macrophage Polarization.
HSPA12A Is a Novel Player in Nonalcoholic Steatohepatitis via Promoting Nuclear PKM2-Mediated M1 Macrophage Polarization.
Diabetes (2018-11-21)
Qiuyue Kong, Nan Li, Hao Cheng, Xiaojin Zhang, Xiaofei Cao, Tao Qi, Leyang Dai, Zhihong Zhang, Xuan Chen, Chuanfu Li, Yuehua Li, Bin Xue, Lei Fang, Li Liu, Zhengnian Ding
ABSTRACT
Nonalcoholic steatohepatitis (NASH) is the most prevalent cause of chronic liver disease worldwide. Macrophage-mediated inflammation plays a critical role in NASH pathogenesis; however, optimum therapies for macrophage activation and NASH remain elusive. HSPA12A encodes a novel member of the HSP70 family. Here, we report that NASH patients showed increased hepatic HSPA12A expression and serum HSPA12A contents. Intriguingly, knockout of HSPA12A (Hspa12a-/- ) in mice attenuated high-fat diet (HFD)-induced hepatic steatosis and injury. HFD-induced macrophage polarization toward an M1 phenotype and inflammatory responses in the liver of Hspa12a-/- mice were also attenuated. Loss- and gain-of-function studies revealed that the de novo lipogenesis in hepatocytes was regulated by the paracrine effects of macrophage HSPA12A rather than by hepatocyte HSPA12A. In-depth molecular analysis revealed that HSPA12A interacted with the M2 isoform of pyruvate kinase (PKM2) in macrophages and increased its nuclear translocation, thereby promoting M1 polarization and secretion of proinflammatory M1 cytokines; this led, ultimately, to hepatocyte steatosis via paracrine effects. Taken together, these findings show that HSPA12A acts as a novel regulator of M1 macrophage polarization and NASH pathogenesis by increasing nuclear PKM2. Strategies that inhibit macrophage HSPA12A might be a potential therapeutic intervention for NASH.
Sigma-Aldrich
DASA-58, ≥98% (HPLC)