Merck
  • Asymmetric 1,2-reduction of enones with potassium borohydride catalyzed by chiral N,N'-dioxide-scandium(III) complexes.
Asymmetric 1,2-reduction of enones with potassium borohydride catalyzed by chiral N,N'-dioxide-scandium(III) complexes.
Organic letters (2012-09-28)
Peng He, Xiaohua Liu, Haifeng Zheng, Wei Li, Lili Lin, Xiaoming Feng
ABSTRACT
The first catalytic enantioselective 1,2-reduction of enones with 0.45 mol equiv potassium borohydride solution catalyzed by a chiral N,N'-dioxide-Sc(III) complex catalyst was accomplished under mild reaction conditions. A number of optically active allylic alcohols were obtained in good to excellent enantioselectivities (up to 95% ee) with nearly quantitative yields.
Sigma-Aldrich
Potassium borohydride, ≥97%