Merck
Selective Functionalization of Aminoheterocycles by a Pyrylium Salt.
Angewandte Chemie (International ed. in English) (2018-07-04)
Daniel Moser, Yaya Duan, Feng Wang, Yuanhong Ma, Matthew J O'Neill, Josep Cornella
ABSTRACT
The functionalization of aminoheterocycles by using a pyrylium tetrafluoroborate reagent (Pyry-BF4 ) is presented. This reagent efficiently condenses with a great variety of heterocyclic amines and primes the C-N bond for nucleophilic aromatic substitution. More than 60 examples for the formation of C-O, C-N, C-S, or C-SO2 R bonds are disclosed herein. In contrast to C-N activation through diazotization and polyalkylation, this method is characterized by its mild conditions and impressive functional-group tolerance. In addition to small-molecule derivatization, Pyry-BF4 allows the introduction of functional groups in a late-stage fashion to furnish highly functionalized structures.
Sigma-Aldrich
Pyrylium tetrafluoroborate, ≥95%