Merck
  • Small molecule promotes β-catenin citrullination and inhibits Wnt signaling in cancer.
Small molecule promotes β-catenin citrullination and inhibits Wnt signaling in cancer.
Nature chemical biology (2017-10-31)
Yi Qu, Jan Roger Olsen, Xing Yuan, Phil F Cheng, Mitchell P Levesque, Karl A Brokstad, Paul S Hoffman, Anne Margrete Oyan, Weidong Zhang, Karl-Henning Kalland, Xisong Ke
ABSTRACT
Wnt (wingless)/β-catenin signaling is critical for tumor progression and is frequently activated in colorectal cancer as a result of the mutation of adenomatous polyposis coli (APC); however, therapeutic agents targeting this pathway for clinical use are lacking. Here we report that nitazoxanide (NTZ), a clinically approved antiparasitic drug, efficiently inhibits Wnt signaling independent of APC. Using chemoproteomic approaches, we have identified peptidyl arginine deiminase 2 (PAD2) as the functional target of NTZ in Wnt inhibition. By targeting PAD2, NTZ increased the deamination (citrullination) and turnover of β-catenin in colon cancer cells. Replacement of arginine residues disrupted the transcriptional activity, and NTZ induced degradation of β-catenin. In Wnt-activated colon cancer cells, knockout of either PAD2 or β-catenin substantially increased resistance to NTZ treatment. Our data highlight the potential of NTZ as a modulator of β-catenin citrullination for the treatment of cancer patients with Wnt pathway mutations.