Merck
  • Role of Surface Chemistry in the Superhydrophobicity of the Springtail Orchesella cincta (Insecta:Collembola).
Role of Surface Chemistry in the Superhydrophobicity of the Springtail Orchesella cincta (Insecta:Collembola).
ACS applied materials & interfaces (2020-02-11)
Lars Schmüser, Wen Zhang, Michael Thomas Marx, Noemi Encinas, Doris Vollmer, Stanislav Gorb, Joe E Baio, Hans Joachim Räder, Tobias Weidner
ABSTRACT
Collembola are ancient arthropods living in soil with extensive exposure to dirt, bacteria, and fungi. To protect from the harsh environmental conditions and to retain a layer of air for breathing when submerged in water, they have evolved a superhydrophobic, liquid-repelling cuticle surface. The nonfouling and self-cleaning properties of springtail cuticle make it an interesting target of biomimetic materials design. Recent research has mainly focused on the intricate microstructures at the cuticle surface. Here we study the role of the cuticle chemistry for the Collembola species Orchesella cincta (Collembola, Entomobryidae). O. cincta uses a relatively simple cuticle structure with primary granules arranged to function as plastrons. In contrast to the Collembolan cuticle featuring structures on multiple length scales that is functional irrespective of surface chemistry, we found that the O. cincta cuticle loses its hydrophobic properties after being rinsed with dichloromethane. Sum frequency generation spectroscopy and time-of-flight secondary ion mass spectrometry in combination with high-resolution mass spectrometry show that a nanometer thin triacylglycerol-containing wax layer at the cuticle surface is essential for maintaining the antiwetting properties. Removal of the wax layer exposes chitin, terpenes, and lipid layers in the cuticle. With respect to biomimetic applications, the results show that, combined with a carefully chosen surface chemistry, superhydrophobicity may be achieved using a relatively unsophisticated surface structure rather than a complex, re-entrant surface structure alone.
Sigma-Aldrich
2,4-Dihydroxybenzophenone, 99%
Pricing and availability is not currently available.