Merck
  • Preparation of Fe3O4 nanoparticle enclosure hydroxylated multi-walled carbon nanotubes for the determination of aconitines in human serum samples.
Preparation of Fe3O4 nanoparticle enclosure hydroxylated multi-walled carbon nanotubes for the determination of aconitines in human serum samples.
Analytica chimica acta (2012-04-10)
Hong-Fei Zhang, Yan-Ping Shi
ABSTRACT
A magnetic carbon nanomaterial for Fe(3)O(4) enclosure hydroxylated multi-walled carbon nanotubes (Fe(3)O(4)-EC-MWCNTs-OH) was prepared by the aggregating effect of Fe(3)O(4) nanoparticle on MWCNTs-OH, and combined with high-performance liquid chromatography (HPLC)/diode array detection (DAD) to determine the aconitines (aconitine, hypaconitine and mesaconitine) in human serum samples. Compared with other extraction modes investigated in experiment, Fe(3)O(4)-EC-MWCNTs-OH sorbents showed a good affinity to target analytes. Some important parameters that could influence extraction efficiency of aconitines, including the extraction mode, amounts of Fe(3)O(4)-EC-MWCNTs-OH, pH of sample solution, extraction time, desorption solvent and desorption time, were optimized. Under optimal conditions, the recoveries of spiked serum samples were between 98.0% and 103.0%; relative standard deviations (RSDs) ranged from 0.9% to 6.2%. The correlation coefficients varied from 0.9996 to 0.9998. The limits of detection ranged from 3.1 ng mL(-1) to 4.1 ng mL(-1) at a signal-to-noise ratio of 3. The experimental results showed that the proposed method was feasible for the analysis of aconitines in serum samples.
Sigma-Aldrich
Aconitine, ≥95% (HPLC), crystalline
Pricing and availability is not currently available.