Merck
Tumour exosome integrins determine organotropic metastasis.
Nature (2015-11-03)
Ayuko Hoshino, Bruno Costa-Silva, Tang-Long Shen, Goncalo Rodrigues, Ayako Hashimoto, Milica Tesic Mark, Henrik Molina, Shinji Kohsaka, Angela Di Giannatale, Sophia Ceder, Swarnima Singh, Caitlin Williams, Nadine Soplop, Kunihiro Uryu, Lindsay Pharmer, Tari King, Linda Bojmar, Alexander E Davies, Yonathan Ararso, Tuo Zhang, Haiying Zhang, Jonathan Hernandez, Joshua M Weiss, Vanessa D Dumont-Cole, Kimberly Kramer, Leonard H Wexler, Aru Narendran, Gary K Schwartz, John H Healey, Per Sandstrom, Knut Jørgen Labori, Elin H Kure, Paul M Grandgenett, Michael A Hollingsworth, Maria de Sousa, Sukhwinder Kaur, Maneesh Jain, Kavita Mallya, Surinder K Batra, William R Jarnagin, Mary S Brady, Oystein Fodstad, Volkmar Muller, Klaus Pantel, Andy J Minn, Mina J Bissell, Benjamin A Garcia, Yibin Kang, Vinagolu K Rajasekhar, Cyrus M Ghajar, Irina Matei, Hector Peinado, Jacqueline Bromberg, David Lyden
ABSTRACT
Ever since Stephen Paget's 1889 hypothesis, metastatic organotropism has remained one of cancer's greatest mysteries. Here we demonstrate that exosomes from mouse and human lung-, liver- and brain-tropic tumour cells fuse preferentially with resident cells at their predicted destination, namely lung fibroblasts and epithelial cells, liver Kupffer cells and brain endothelial cells. We show that tumour-derived exosomes uptaken by organ-specific cells prepare the pre-metastatic niche. Treatment with exosomes from lung-tropic models redirected the metastasis of bone-tropic tumour cells. Exosome proteomics revealed distinct integrin expression patterns, in which the exosomal integrins α6β4 and α6β1 were associated with lung metastasis, while exosomal integrin αvβ5 was linked to liver metastasis. Targeting the integrins α6β4 and αvβ5 decreased exosome uptake, as well as lung and liver metastasis, respectively. We demonstrate that exosome integrin uptake by resident cells activates Src phosphorylation and pro-inflammatory S100 gene expression. Finally, our clinical data indicate that exosomal integrins could be used to predict organ-specific metastasis.
Sigma-Aldrich
Anti-Integrin beta-3 Antibody, from rabbit, purified by affinity chromatography
Pricing and availability is not currently available.