Merck
  • Mechanistic Effects of Spironolactone on Cardiovascular and Renal Biomarkers in Heart Failure With Preserved Ejection Fraction: A TOPCAT Biorepository Study.
Mechanistic Effects of Spironolactone on Cardiovascular and Renal Biomarkers in Heart Failure With Preserved Ejection Fraction: A TOPCAT Biorepository Study.
Circulation. Heart failure (2020-01-21)
Peder L Myhre, Muthiah Vaduganathan, Eileen O'Meara, Brian L Claggett, Simon de Denus, Petr Jarolim, Inder S Anand, Bertram Pitt, Jean L Rouleau, Scott D Solomon, Marc A Pfeffer, Akshay S Desai
ABSTRACT
Spironolactone has been demonstrated to reduce heart failure (HF) hospitalization in patients with HF with preserved ejection fraction in the Americas region of the TOPCAT trial (Treatment of Preserved Cardiac Function Heart Failure With an Aldosterone Antagonist Trial). We assessed effects of 12 months of treatment with spironolactone on biomarkers reflecting myocardial stress, myocardial injury, renal function, and systemic inflammation. This TOPCAT biorepository substudy evaluated 247 patients (14% of the total 1767 patients in the Americas region) with symptomatic HF, ejection fraction ≥45%, and elevated natriuretic peptides or a prior HF hospitalization. Paired blood samples at baseline and after 12 months of treatment with spironolactone or placebo were available in 204 patients. At baseline, the median (interquartile range) concentration of BNP (B-type natriuretic peptide) was 124 (69-197) ng/L, NT-proBNP (N-terminal-pro-B-type natriuretic peptide) 624 (307-1312) ng/L, hs-cTnI (high sensitivity cardiac troponin I) 6.3 (3.4-13.0) ng/L, hs-CRP (high sensitivity C-reactive protein) 2.8 (1.3-6.1) mg/L, uric acid 7.2 (5.8-8.7) mg/dL, and urine protein-creatinine ratio 0.11 (0.08-0.20) mg/mg. Compared with placebo-assigned participants at 12 months, those randomized to spironolactone experienced greater reductions from baseline in levels of NT-proBNP (P=0.017) and BNP (P=0.002); these differences persisted after adjustment for demographics, comorbidities, estimated glomerular filtration rate, and enrollment strata. No between-group differences in changes in hs-cTnI, CRP, uric acid, or urine protein-creatinine ratio were observed. This TOPCAT biorepository substudy suggests potential effects on markers of cardiac wall stress or filling pressures during 12 months of treatment with spironolactone in patients with chronic HF with preserved ejection fraction. URL: https://www.clinicaltrials.gov. Unique identifier: NCT00094302.
Sigma-Aldrich
(+)-Biotin 4-nitrophenyl ester, 98%
Pricing and availability is not currently available.