Merck
  • Cilia-driven asymmetric Hedgehog signalling determines the amphioxus left-right axis by controlling Dand5 expression.
Cilia-driven asymmetric Hedgehog signalling determines the amphioxus left-right axis by controlling Dand5 expression.
Development (Cambridge, England) (2019-12-13)
Xin Zhu, Chenggang Shi, Yanhong Zhong, Xian Liu, Qiuning Yan, Xiaotong Wu, Yiquan Wang, Guang Li
ABSTRACT
Cilia rotation-driven nodal flow is crucial for the left-right (L-R) break in symmetry in most vertebrates. However, the mechanism by which the flow signal is translated to asymmetric gene expression has been insufficiently addressed. Here, we show that Hedgehog (Hh) signalling is asymmetrically activated (L<R) in the region in which initial asymmetric Dand5 expression is detected. Upregulation of Hh signalling on the left side of wild-type embryos induces ectopic Dand5 expression on the left side, and the unilateral recovery of Hh signalling in Hh homozygous mutants induces Dand5 expression in the Hh signal recovery side. Immunofluorescence analysis results revealed that Hh fusion protein is asymmetrically enriched in the anterior-right paraxial mesoderm at the early neurula stage. Inhibiting embryonic cilia motility using methylcellulose (MC) blocks Hh protein enrichment on the right hand side and randomizes Dand5 expression and organ positioning along the L-R axis. These findings present a model showing that cilia movement is crucial for the symmetry breaks in amphioxus through asymmetric Hh protein transport. The resultant asymmetric Hh signalling provides a clue into the induction of asymmetric Dand5 expression.This article has an associated 'The people behind the papers' interview.
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture