Merck
  • Expressions of TGF-β2, bFGF and ICAM-1 in lens epithelial cells of complicated cataract with silicone oil tamponade.
Expressions of TGF-β2, bFGF and ICAM-1 in lens epithelial cells of complicated cataract with silicone oil tamponade.
International journal of ophthalmology (2017-07-22)
Bei Liu, Jing Gao, Bo-Chang Lyu, Shan-Shuang Du, Cheng Pei, Zhong-Qiao Zhu, Bo Ma
ABSTRACT
To investigate the expression differences of transforming growth factor-β2 (TGF-β2), basic fibroblast growth factor (bFGF) and intercellular cell-adhesion molecule-1 (ICAM-1) in lens epithelial cells (LECs) of complicated cataract with silicone oil tamponade and age-related cataract. Totally 150 eyes of 150 patients (aged 35 to 77y) were investigated, including 75 patients with complicated cataract after silicone oil tamponade and 75 patients with age-related cataract. The central piece of anterior capsules was collected during cataract surgery. TGF-β2, bFGF and ICAM-1 were detected in the 60 specimens of the two groups by immunohistochemistry. The expression levels of the three kinds of messenger ribonucleic acid (mRNA) were determined by real-time quantitative reverse transcription-polymerase chain reaction in the 90 specimens of the two groups. TGF-β2 was detected in the cytomembrane and cytoplasm of the LECs and bFGF was detected in the nucleus. ICAM-1 was positive in the cytomembrane of the LECs and the distribution of positive cells was uneven. The mRNA genes expression of the TGF-β2, bFGF and ICAM-1 was significant differences between the two groups and markedly increased in complicated cataract group (P<0.05). The up-regulated TGF-β2, bFGF and ICAM-1 maybe associate with the occurrence and development of complicated cataract with silicone oil tamponade.
Sigma-Aldrich
Donkey Anti-Mouse IgG Antibody, biotin conjugate, Species Adsorbed, 0.8 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
Donkey Anti-Rabbit IgG Antibody, biotin-SP conjugate, Species Adsorbed, Chemicon®, from donkey