Merck
  • Overexpression of microRNA-9 inhibits 3T3-L1 cell adipogenesis by targeting PNPLA3 via activation of AMPK.
Overexpression of microRNA-9 inhibits 3T3-L1 cell adipogenesis by targeting PNPLA3 via activation of AMPK.
Gene (2019-11-25)
Beibei Xu, Jianhua Shen, Dan Li, Baoshuo Ning, Liping Guo, Hao Bing, Jiayu Chen, Yiling Li
ABSTRACT
Adipocyte differentiation, which plays an important role in the development of obesity, involves complex molecular networks in which microRNAs (miRNAs) are essential. Here, we show that miR-9 expression was upregulated during adipogenesis in 3T3-L1 cells. miR-9 overexpression reduced the accumulation of lipid droplets and the content of triglycerides by downregulating patatin-like phospholipase domain-containing protein 3 (PNPLA3). PNPLA3 knockdown or miR-9 overexpression downregulated the protein expression of peroxisome proliferator-activated receptor γ, oxidative tissue-enrichedPAT, and LC3, and promoted the phosphorylation of AMP-activated protein kinase (AMPK), which was inhibited by treatment with the AMPK inhibitor compound C. These results indicate that miR-9 inhibits adipocyte differentiation by targeting PNPLA3 through the AMPK pathway.
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human PNPLA3
Pricing and availability is not currently available.