Merck
  • Dysregulation of autophagy and stress granule-related proteins in stress-driven Tau pathology.
Dysregulation of autophagy and stress granule-related proteins in stress-driven Tau pathology.
Cell death and differentiation (2018-11-18)
Joana Margarida Silva, Sara Rodrigues, Belém Sampaio-Marques, Patrícia Gomes, Andreia Neves-Carvalho, Chrysoula Dioli, Carina Soares-Cunha, Brandon F Mazuik, Akihiko Takashima, Paula Ludovico, Benjamin Wolozin, Nuno Sousa, Ioannis Sotiropoulos
ABSTRACT
Imbalance of neuronal proteostasis associated with misfolding and aggregation of Tau protein is a common neurodegenerative feature in Alzheimer's disease (AD) and other Tauopathies. Consistent with suggestions that lifetime stress may be an important AD precipitating factor, we previously reported that environmental stress and high glucocorticoid (GC) levels induce accumulation of aggregated Tau; however, the molecular mechanisms for such process remain unclear. Herein, we monitor a novel interplay between RNA-binding proteins (RBPs) and autophagic machinery in the underlying mechanisms through which chronic stress and high GC levels impact on Tau proteostasis precipitating Tau aggregation. Using molecular, pharmacological and behavioral analysis, we demonstrate that chronic stress and high GC trigger mTOR-dependent inhibition of autophagy, leading to accumulation of Tau aggregates and cell death in P301L-Tau expressing mice and cells. In parallel, we found that environmental stress and GC disturb cellular homeostasis and trigger the insoluble accumulation of different RBPs, such as PABP, G3BP1, TIA-1, and FUS, shown to form stress granules (SGs) and Tau aggregation. Interestingly, an mTOR-driven pharmacological stimulation of autophagy attenuates the GC-driven accumulation of Tau and SG-related proteins as well as the related cell death, suggesting a critical interface between autophagy and the response of the SG-related protein in the neurodegenerative potential of chronic stress and GC. These studies provide novel insights into the RNA-protein intracellular signaling regulating the precipitating role of environmental stress and GC on Tau-driven brain pathology.
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-acetyl Cortactin Antibody, from rabbit, purified by affinity chromatography
Pricing and availability is not currently available.
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.