Merck
  • Differentiation and characterization of human pluripotent stem cell-derived brain microvascular endothelial cells.
Differentiation and characterization of human pluripotent stem cell-derived brain microvascular endothelial cells.
Methods (San Diego, Calif.) (2015-11-01)
Matthew J Stebbins, Hannah K Wilson, Scott G Canfield, Tongcheng Qian, Sean P Palecek, Eric V Shusta
ABSTRACT
The blood-brain barrier (BBB) is a critical component of the central nervous system (CNS) that regulates the flux of material between the blood and the brain. Because of its barrier properties, the BBB creates a bottleneck to CNS drug delivery. Human in vitro BBB models offer a potential tool to screen pharmaceutical libraries for CNS penetration as well as for BBB modulators in development and disease, yet primary and immortalized models respectively lack scalability and robust phenotypes. Recently, in vitro BBB models derived from human pluripotent stem cells (hPSCs) have helped overcome these challenges by providing a scalable and renewable source of human brain microvascular endothelial cells (BMECs). We have demonstrated that hPSC-derived BMECs exhibit robust structural and functional characteristics reminiscent of the in vivo BBB. Here, we provide a detailed description of the methods required to differentiate and functionally characterize hPSC-derived BMECs to facilitate their widespread use in downstream applications.
Sigma-Aldrich
Anti-BCRP1 Antibody, clone 5D3, clone 5D3, Chemicon®, from mouse