Merck
  • Prenatal PPARα activation by clofibrate increases subcutaneous fat browning in male C57BL/6J mice fed a high-fat diet during adulthood.
Prenatal PPARα activation by clofibrate increases subcutaneous fat browning in male C57BL/6J mice fed a high-fat diet during adulthood.
PloS one (2017-11-03)
Szu-Han Chen, Pei-Min Chao
ABSTRACT
We tested the hypothesis that prenatal administration of PPARα agonist clofibrate may permanently increase browning capacity of developing white adipose tissue (WAT). Pregnant C57BL/6J mice were fed a basal diet, without (C) or with 0.5% clofibrate (CF, a PPARα agonist) throughout pregnancy. After parturition, only male offspring were used; all suckled their mothers (who were eating the C diet) and after weaning, they ate a standard chow diet for 4 wk, followed by a high-fat diet (HFD) for 5 wk. Administration of CF up-regulated serum concentrations and hepatic expression of FGF21 in fetuses, with a return to basal levels after CF withdrawal. At postnatal day 84 (P84), CF-offspring had significantly higher expression of thermogenic genes (Ucp1, Cidea, Ppara Ppargc1a, Cpt1b) and UCP1 protein levels in response to HFD in inguinal fat, but not in retroperitoneal (combined with perirenal) or epididymal fat. Based on UCP1 levels in inguinal fat on P7, P14, and P21, appearance of the transient brown-adipocyte phenotype seemed to be hastened by CF exposure. We concluded that giving CF to pregnant mice programmed greater HFD-induced WAT browning in subcutaneous, but not in visceral fat, in their male offspring at adulthood.
Sigma-Aldrich
Clofibrate, liquid
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, clone LNC1, ascites fluid, clone LNC1, Chemicon®