Merck
Cardiac mitochondrial connexin 43 regulates apoptosis.
Biochemical and biophysical research communications (2006-11-17)
Farida Goubaeva, Maya Mikami, Sarah Giardina, Bo Ding, Junichi Abe, Jay Yang
ABSTRACT
Connexin 43 (Cx43) is thought to be present largely in the plasma membrane and its function solely to provide low resistance electrical connection between myocytes. A recent report suggested the presence of Cx43 in the mitochondria as well. We confirmed the presence of Cx43 in the mitochondria isolated from adult rat ventricles with the Cx43 immunoreactivity fractionating to the outer mitochondrial membrane. Mitochondrial Cx43 is mostly phosphorylated only detected by a phospho-specific antibody. Using a Ca2+ -sensitive electrode and Western blot, we showed that the gap junction inhibitors 18-beta-glycyrrhetinic acid (beta-GA), oleamide, and heptanol all induced concomitant release of Ca2+ and cytochrome C in isolated mitochondria whereas the inactive analog 18-beta-glycyrrhizic acid failed to do so. In low density neonatal myocyte culture with no appreciable cell-cell contacts, beta-GA induced apoptosis as assessed by TUNEL staining. Our results suggest a novel role of Cx43 as a regulator of mitochondrial physiology and myocyte apoptosis.
Sigma-Aldrich
Anti-Connexin 43 Antibody, clone 4E6.2, clone 4E6.2, Chemicon®, from mouse
Pricing and availability is not currently available.