- Cocaine-evoked negative symptoms require AMPA receptor trafficking in the lateral habenula.
Cocaine-evoked negative symptoms require AMPA receptor trafficking in the lateral habenula.
Nature neuroscience (2015-02-03)
Frank J Meye, Kristina Valentinova, Salvatore Lecca, Lucile Marion-Poll, Matthieu J Maroteaux, Stefano Musardo, Imane Moutkine, Fabrizio Gardoni, Richard L Huganir, François Georges, Manuel Mameli
PMID25643299
ABSTRACT
Addictive substances mediate positive and negative states promoting persistent drug use. However, substrates for aversive effects of drugs remain elusive. We found that, in mouse lateral habenula (LHb) neurons targeting the rostromedial tegmental nucleus, cocaine enhanced glutamatergic transmission, reduced K(+) currents and increased excitability. GluA1 trafficking in LHb was instrumental for these cocaine-evoked modifications and drug-driven aversive behaviors. Altogether, our results suggest that long-lasting adaptations in LHb shape negative symptoms after drug taking.
Sigma-Aldrich
Anti-phospho-GluR1 (Ser831) Antibody, clone N453, rabbit monoclonal, culture supernatant, clone N453, Upstate®